2,4-Dichloro-3,5-difluoroaniline 97%

Lieferant: Apollo Scientific

PC902646-1G PC902646-25G PC902646-5G
APOSPC902646-1GEA 125 EUR
APOSPC902646-1G APOSPC902646-25G APOSPC902646-5G
2,4-Dichloro-3,5-difluoroaniline 97%
2,4-Dichloro-3,5-difluoroaniline
Formel: C₆H₃Cl₂F₂N
Molecular Weight: 197.998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827974
CAS-Nummer: 50408-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR