4-Acetamido-3-fluorobenzoic acid 97%

Lieferant: Apollo Scientific

PC902663-1G PC902663-250MG
APOSPC902663-1GEA 531 EUR
APOSPC902663-1G APOSPC902663-250MG
4-Acetamido-3-fluorobenzoic acid 97%
4-Acetamido-3-fluorobenzoic acid
Formel: C₉H₈FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00215837
CAS-Nummer: 713-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR