2-(2-Fluoroethoxy)aniline 97%

Lieferant: Apollo Scientific

PC902757-1G PC902757-5G
APOSPC902757-1GEA 219 EUR
APOSPC902757-1G APOSPC902757-5G
2-(2-Fluoroethoxy)aniline 97%
2-(2-Fluoroethoxy)aniline
Formel: C₈H₁₀FNO
Molecular Weight: 155,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909942
CAS-Nummer: 1547-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR