2-Methyl-3,4,6-trifluorobenzoic acid 95%

Lieferant: Apollo Scientific

PC902765-1G PC902765-250MG
APOSPC902765-1GEA 415 EUR
APOSPC902765-1G APOSPC902765-250MG
2-Methyl-3,4,6-trifluorobenzoic acid 95%
2-Methyl-3,4,6-trifluorobenzoic acid
Formel: C₈H₅F₃O₂
Molecular Weight: 190.119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410917
CAS-Nummer: 119916-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR