(S)-2,2-Dimethyl-1-trifluoromethyl-propylamine 95%

Lieferant: Apollo Scientific

PC902868-1G PC902868-250MG
APOSPC902868-1GEA 712 EUR
APOSPC902868-1G APOSPC902868-250MG
(S)-2,2-Dimethyl-1-trifluoromethyl-propylamine 95%
(S)-2,2-Dimethyl-1-trifluoromethyl-propylamine
Formel: C₆H₁₂F₃N
Molecular Weight: 155,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08058822
CAS-Nummer: 736127-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR