1,3-Difluoro-5-methylsulphonylbenzene 98%

Lieferant: Apollo Scientific

Synonyme: 1,3-Difluoro-5-(methylsulphonyl)benzene

PC902949-1G PC902949-5G
APOSPC902949-1GEA 203 EUR
APOSPC902949-1G APOSPC902949-5G
1,3-Difluoro-5-methylsulphonylbenzene 98%
1,3-Difluoro-5-methylsulphonylbenzene
Formel: C₇H₆F₂O₂S
Molecular Weight: 192,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09972174
CAS-Nummer: 171421-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR