3-Bromo-5-chloro-2-fluorobenzeneboronic acid 95%

Lieferant: Apollo Scientific

PC903364-10G PC903364-1G PC903364-5G
APOSPC903364-10GEA 654 EUR
APOSPC903364-10G APOSPC903364-1G APOSPC903364-5G
3-Bromo-5-chloro-2-fluorobenzeneboronic acid 95%
3-Bromo-5-chloro-2-fluorobenzeneboronic acid
Formel: C₆H₄BBrClFO₂
Molecular Weight: 253,261 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28143924
CAS-Nummer: 2377608-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR