1-(3-Fluorophenyl)cyclohexanol 95%

Lieferant: Apollo Scientific

PC903632-1G PC903632-250MG
APOSPC903632-1GEA 45.9 EUR
APOSPC903632-1G APOSPC903632-250MG
1-(3-Fluorophenyl)cyclohexanol 95%
1-(3-Fluorophenyl)cyclohexanol
Formel: C₁₂H₁₅FO
Lagertemperatur: Raumtemperatur
MDL: MFCD00218768
CAS-Nummer: 1496-35-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR