Methyl 2-amino-3,5-difluorobenzoate 95%

Lieferant: Apollo Scientific

PC903644-1G PC903644-5G
APOSPC903644-1GEA 55.1 EUR
APOSPC903644-1G APOSPC903644-5G
Methyl 2-amino-3,5-difluorobenzoate 95%
Methyl 2-amino-3,5-difluorobenzoate
Formel: C₈H₇F₂NO₂
Molecular Weight: 187,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12159866
CAS-Nummer: 1184559-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR