2-Methyl-4-(trifluoromethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: [2-Methyl-4-(trifluoromethyl)phenyl]methanol

PC903778-1G PC903778-5G
APOSPC903778-1GEA 429 EUR
APOSPC903778-1G APOSPC903778-5G
2-Methyl-4-(trifluoromethyl)benzyl alcohol 97%
2-Methyl-4-(trifluoromethyl)benzyl alcohol
Formel: C₉H₉F₃O
Molecular Weight: 190.162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18399874
CAS-Nummer: 888739-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR