4-Chloro-6-fluoro-1H-indole 95+%

Lieferant: Apollo Scientific
Achtung

PC903779-100MG PC903779-250MG
APOSPC903779-100MGEA 58 EUR
APOSPC903779-100MG APOSPC903779-250MG
4-Chloro-6-fluoro-1H-indole 95+%
4-Chloro-6-fluoro-1H-indole
Formel: C₈H₅ClFN
Molecular Weight: 169,583 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07781557
CAS-Nummer: 885520-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR