1-Chloro-2-fluoro-4-methyl-5-nitrobenzene 99%

Lieferant: Apollo Scientific
Achtung

PC903922-1G PC903922-25G PC903922-5G
APOSPC903922-1GEA 21.9 EUR
APOSPC903922-1G APOSPC903922-25G APOSPC903922-5G
1-Chloro-2-fluoro-4-methyl-5-nitrobenzene 99%
1-Chloro-2-fluoro-4-methyl-5-nitrobenzene
Formel: C₇H₅ClFNO₂
Molecular Weight: 189,572 g/mol
Schmelzpunkt: 59…60 °C
Lagertemperatur: Kühlschrank
MDL: MFCD15527686
CAS-Nummer: 118664-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR