Methyl 4-chloro-5-fluoroanthranilate 95%
Lieferant: Apollo Scientific
PC903975-10G
PC903975-1G
PC903975-5G
APOSPC903975-10GEA
1130
EUR
APOSPC903975-10G
APOSPC903975-1G
APOSPC903975-5G
Methyl 4-chloro-5-fluoroanthranilate 95%
Methyl 4-chloro-5-fluoroanthranilate
Formel:
C₈H₇ClFNO₂ Molecular Weight: 203,598 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15523631 CAS-Nummer: 104901-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...