(S)-4,4-Difluoropyrrolidine-2-carboxylic acid hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-4,4-DiF-Pro-OH·HCl

PC903976-100MG PC903976-250MG
APOSPC903976-100MGEA 49.3 EUR
APOSPC903976-100MG APOSPC903976-250MG
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid hydrochloride 95+%
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid hydrochloride
Formel: C₅H₇F₂NO₂·ClH
Molecular Weight: 187,572 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22199978
CAS-Nummer: 1588480-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR