Potassium 2-(tetrahydro-2h-pyran-2-yloxy)ethyltrifluoroborate

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium trifluoro-[2-(oxan-2-yloxy)ethyl]boranuide, Potassium trifluoro(2-((tetrahydro-2H-pyran-2-yl)oxy)ethyl)borate

PC904235-1G PC904235-250MG PC904235-5G
APOSPC904235-1GEA 89.9 EUR
APOSPC904235-1G APOSPC904235-250MG APOSPC904235-5G
Potassium 2-(tetrahydro-2h-pyran-2-yloxy)ethyltrifluoroborate
Potassium 2-(tetrahydro-2h-pyran-2-yloxy)ethyltrifluoroborate
Formel: C₇H₁₃BF₃O₂·K
Molecular Weight: 236,081 g/mol
Schmelzpunkt: 182…185 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD22684702
CAS-Nummer: 1408168-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR