1-(2-Amino-5-chloro-4-fluoro-phenyl)ethanone 95%

Lieferant: Apollo Scientific

PC904396-1G PC904396-250MG
APOSPC904396-1GEA 719 EUR
APOSPC904396-1G APOSPC904396-250MG
1-(2-Amino-5-chloro-4-fluoro-phenyl)ethanone 95%
1-(2-Amino-5-chloro-4-fluoro-phenyl)ethanone
Formel: C₈H₇ClFNO
Molecular Weight: 187,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11113354
CAS-Nummer: 937816-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR