2-Fluoro-5-(methylsulphonyl)toluene 95%

Lieferant: Apollo Scientific

PC904459-1G PC904459-250MG
APOSPC904459-1GEA 126 EUR
APOSPC904459-1G APOSPC904459-250MG
2-Fluoro-5-(methylsulphonyl)toluene 95%
2-Fluoro-5-(methylsulphonyl)toluene
Formel: C₈H₉FO₂S
Molecular Weight: 188,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527249
CAS-Nummer: 828270-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR