5-(2-Fluorophenyl)-2-piperidone 95%

Lieferant: Apollo Scientific

PC904484-1G PC904484-250MG
APOSPC904484-1GEA 734 EUR
APOSPC904484-1G APOSPC904484-250MG
5-(2-Fluorophenyl)-2-piperidone 95%
5-(2-Fluorophenyl)-2-piperidone
Formel: C₁₁H₁₂FNO
Molecular Weight: 193,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19348296
CAS-Nummer: 80942-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR