(S)-3-Fluoropyrrolidine 96%

Lieferant: Apollo Scientific
Achtung

PC904564-1G PC904564-5G
APOSPC904564-1GEA 310 EUR
APOSPC904564-1G APOSPC904564-5G
(S)-3-Fluoropyrrolidine 96%
(S)-3-Fluoropyrrolidine
Formel: C₄H₈FN
Molecular Weight: 89,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738461
CAS-Nummer: 136725-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR