1-(2,2,2-Trifluoroethyl)piperidin-4-one 98%

Lieferant: Apollo Scientific

PC904625-100MG PC904625-1G PC904625-250MG
APOSPC904625-100MGEA 271 EUR
APOSPC904625-100MG APOSPC904625-1G APOSPC904625-250MG
1-(2,2,2-Trifluoroethyl)piperidin-4-one 98%
1-(2,2,2-Trifluoroethyl)piperidin-4-one
Formel: C₇H₁₀F₃NO
Molecular Weight: 181.156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09936907
CAS-Nummer: 81363-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR