2-(Trifluoromethyl)-1H-imidazole-5-carboxylic acid 96%

Lieferant: Apollo Scientific

PC904733-1G PC904733-5G
APOSPC904733-1GEA 0 EUR
APOSPC904733-1G APOSPC904733-5G
2-(Trifluoromethyl)-1H-imidazole-5-carboxylic acid 96%
2-(Trifluoromethyl)-1H-imidazole-5-carboxylic acid
Formel: C₅H₃F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10696306
CAS-Nummer: 78016-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR