2,4-Difluoro-3-formylbenzonitrile 98%

Lieferant: Apollo Scientific

PC904756-1G PC904756-5G
APOSPC904756-1GEA 151 EUR
APOSPC904756-1G APOSPC904756-5G
2,4-Difluoro-3-formylbenzonitrile 98%
2,4-Difluoro-3-formylbenzonitrile
Formel: C₈H₃F₂NO
Molecular Weight: 167,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907858
CAS-Nummer: 149489-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR