5-Fluoro-7-azaindole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

PC904839-1G
APOSPC904839-1GEA 71.2 EUR
APOSPC904839-1G
5-Fluoro-7-azaindole-3-carboxylic acid 95%
5-Fluoro-7-azaindole-3-carboxylic acid
Formel: C₈H₅FN₂O₂
Molecular Weight: 180,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12962623
CAS-Nummer: 1067193-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR