5-Amino-4-bromo-2-fluorobenzonitrile 96%

Lieferant: Apollo Scientific

PC904887-10G PC904887-1G PC904887-5G
APOSPC904887-10GEA 672 EUR
APOSPC904887-10G APOSPC904887-1G APOSPC904887-5G
5-Amino-4-bromo-2-fluorobenzonitrile 96%
5-Amino-4-bromo-2-fluorobenzonitrile
Formel: C₇H₄BrFN₂
Molecular Weight: 215.022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977386
CAS-Nummer: 893615-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR