4-Hydroxymethyl-2,2-difluoro-1,3-benzodioxole 96%

Lieferant: Apollo Scientific
Achtung

PC905024-10G PC905024-1G PC905024-5G
APOSPC905024-10GEA 870 EUR
APOSPC905024-10G APOSPC905024-1G APOSPC905024-5G
4-Hydroxymethyl-2,2-difluoro-1,3-benzodioxole 96%
4-Hydroxymethyl-2,2-difluoro-1,3-benzodioxole
Formel: C₈H₆F₂O₃
Molecular Weight: 188.128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202698
CAS-Nummer: 157437-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR