3-Bromo-5-fluoro-4-hydroxybenzaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

PC905038-1G PC905038-5G
APOSPC905038-1GEA 26.1 EUR
APOSPC905038-1G APOSPC905038-5G
3-Bromo-5-fluoro-4-hydroxybenzaldehyde 95+%
3-Bromo-5-fluoro-4-hydroxybenzaldehyde
Formel: C₇H₄BrFO₂
Molecular Weight: 219.008 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21606586
CAS-Nummer: 185345-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR