N-(3-Fluoro-2-methyl-phenyl)-formamide 95%

Lieferant: Apollo Scientific

PC905067-1G PC905067-250MG
APOSPC905067-1GEA 371 EUR
APOSPC905067-1G APOSPC905067-250MG
N-(3-Fluoro-2-methyl-phenyl)-formamide 95%
N-(3-Fluoro-2-methyl-phenyl)-formamide
Formel: C₈H₈FNO
Molecular Weight: 153,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19441851
CAS-Nummer: 153797-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR