4-Fluoro-1-methyl-1H-indazole 95%

Lieferant: Apollo Scientific
Achtung

PC905092-100MG PC905092-1G PC905092-250MG
APOSPC905092-100MGEA 21.9 EUR
APOSPC905092-100MG APOSPC905092-1G APOSPC905092-250MG
4-Fluoro-1-methyl-1H-indazole 95%
4-Fluoro-1-methyl-1H-indazole
Formel: C₈H₇FN₂
Molecular Weight: 150,153 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11052635
CAS-Nummer: 1092961-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR