N-(2-Fluoro-6-methoxyphenyl)formamide 95%

Lieferant: Apollo Scientific

PC905146-1G PC905146-250MG
APOSPC905146-1GEA 647 EUR
APOSPC905146-1G APOSPC905146-250MG
N-(2-Fluoro-6-methoxyphenyl)formamide 95%
N-(2-Fluoro-6-methoxyphenyl)formamide
Formel: C₈H₈FNO₂
Molecular Weight: 169,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19441833
CAS-Nummer: 1313712-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR