2-Chloro-5-(trifluoromethyl)nicotinonitrile 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-(trifluoromethyl)pyridine-3-carbonitrile
PC905310-1G
PC905310-250MG
APOSPC905310-1GEA
243
EUR
APOSPC905310-1G
APOSPC905310-250MG
2-Chloro-5-(trifluoromethyl)nicotinonitrile 98%
2-Chloro-5-(trifluoromethyl)nicotinonitrile
Formel:
C₇H₂ClF₃N₂ Molecular Weight: 206,552 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697685 CAS-Nummer: 624734-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...