3-(Fluoromethyl)aniline 95%
Lieferant: Apollo Scientific
PC905438-1G
PC905438-250MG
APOSPC905438-1GEA
217
EUR
APOSPC905438-1G
APOSPC905438-250MG
3-(Fluoromethyl)aniline 95%
3-(Fluoromethyl)aniline
Formel:
C₇H₈FN Molecular Weight: 125,144 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00466952 CAS-Nummer: 456-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...