2-Cyano-5-fluorobenzaldehyde 95%
Lieferant: Apollo Scientific
Synonyme:
4-Fluoro-2-formylbenzonitrile
PC905464-1G
PC905464-5G
APOSPC905464-1GEA
81.2
EUR
APOSPC905464-1G
APOSPC905464-5G
2-Cyano-5-fluorobenzaldehyde 95%
2-Cyano-5-fluorobenzaldehyde
Formel:
C₈H₄FNO Molecular Weight: 149,122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16619724 CAS-Nummer: 894779-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...