4-Amino-2-fluoro-5-methoxybenzonitrile 98%

Lieferant: Apollo Scientific

PC905561-1G PC905561-5G
APOSPC905561-1GEA 42.2 EUR
APOSPC905561-1G APOSPC905561-5G
4-Amino-2-fluoro-5-methoxybenzonitrile 98%
4-Amino-2-fluoro-5-methoxybenzonitrile
Formel: C₈H₇FN₂O
Molecular Weight: 166,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26407017
CAS-Nummer: 1441723-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR