6-Chloro-5-fluoro-2-oxindole 95%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-5-fluoroindolin-2-one
PC905563-5G
APOSPC905563-5GEA
115
EUR
APOSPC905563-5G
6-Chloro-5-fluoro-2-oxindole 95%
6-Chloro-5-fluoro-2-oxindole
Formel:
C₈H₅ClFNO Molecular Weight: 185,583 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09839325 CAS-Nummer: 100487-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...