4-Amino-2-fluoro-N-methylbenzamide 98%

Lieferant: Apollo Scientific

PC905580-10G PC905580-25G PC905580-100G
APOSPC905580-10GEA 101 EUR
APOSPC905580-10G APOSPC905580-25G APOSPC905580-100G
4-Amino-2-fluoro-N-methylbenzamide 98%
4-Amino-2-fluoro-N-methylbenzamide
Formel: C₈H₉FN₂O
Molecular Weight: 168.168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909322
CAS-Nummer: 915087-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR