3-Bromo-4-chloro-2-fluorobenzyl alcohol 95%

Lieferant: Apollo Scientific

Synonyme: (3-Bromo-4-chloro-2-fluorophenyl)methanol

PC905621-1G PC905621-250MG PC905621-5G
APOSPC905621-1GEA 328 EUR
APOSPC905621-1G APOSPC905621-250MG APOSPC905621-5G
3-Bromo-4-chloro-2-fluorobenzyl alcohol 95%
3-Bromo-4-chloro-2-fluorobenzyl alcohol
Formel: C₇H₅BrClFO
Molecular Weight: 239,469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28130389
CAS-Nummer: 1824057-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR