3-Ethenylazetidine trifluoroacetic acid

Lieferant: Apollo Scientific

PC905643-1G PC905643-5G
APOSPC905643-1GEA 655 EUR
APOSPC905643-1G APOSPC905643-5G
3-Ethenylazetidine trifluoroacetic acid
3-Ethenylazetidine trifluoroacetic acid
Formel: C₅H₉N·CF₃CO₂H
Lagertemperatur: Raumtemperatur
MDL: MFCD28166293
CAS-Nummer: 1630907-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR