4-Bromo-1-(trifluoromethyl)pyrazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-(trifluoromethyl)-1H-pyrazole

PC905979-100MG PC905979-250MG
APOSPC905979-100MGEA 429 EUR
APOSPC905979-100MG APOSPC905979-250MG
4-Bromo-1-(trifluoromethyl)pyrazole 95%
4-Bromo-1-(trifluoromethyl)pyrazole
Formel: C₄H₂BrF₃N₂
Molecular Weight: 214,971 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD27988093
CAS-Nummer: 1046831-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR