2-(Trifluoromethoxy)benzoylacetonitrile 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Oxo-3-[2-(trifluoromethoxy)phenyl]propanenitrile, 3-Oxo-3-[2-(trifluoromethoxy)phenyl]propionitrile
PC9063-1G
PC9063-250MG
PC9063-5G
PC9063-25G
APOSPC9063-1GEA
116
EUR
APOSPC9063-1G
APOSPC9063-250MG
APOSPC9063-5G
APOSPC9063-25G
2-(Trifluoromethoxy)benzoylacetonitrile 98%
2-(Trifluoromethoxy)benzoylacetonitrile
Formel:
C₁₀H₆F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458117 CAS-Nummer: 914636-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...