4-Amino-2-chloro-5-fluorobenzonitrile 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC906546-1G
PC906546-250MG
APOSPC906546-1GEA
155
EUR
APOSPC906546-1G
APOSPC906546-250MG
4-Amino-2-chloro-5-fluorobenzonitrile 95+%
4-Amino-2-chloro-5-fluorobenzonitrile
Formel:
C₇H₄ClFN₂ Molecular Weight: 170,571 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110982 CAS-Nummer: 1228376-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...