2-Fluoro-3-methylbutyric acid 95%

Lieferant: Apollo Scientific

PC906555-100MG PC906555-1G PC906555-250MG
APOSPC906555-100MGEA 417 EUR
APOSPC906555-100MG APOSPC906555-1G APOSPC906555-250MG
2-Fluoro-3-methylbutyric acid 95%
2-Fluoro-3-methylbutyric acid
Formel: C₅H₉FO₂
Molecular Weight: 120,122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01737272
CAS-Nummer: 1578-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR