5-Chloro-6-fluoropyridin-2-amine 95%

Lieferant: Apollo Scientific

PC906713-1G PC906713-250MG
APOSPC906713-1GEA 200 EUR
APOSPC906713-1G APOSPC906713-250MG
5-Chloro-6-fluoropyridin-2-amine 95%
5-Chloro-6-fluoropyridin-2-amine
Formel: C₅H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18253522
CAS-Nummer: 1378595-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR