1-(Trifluoromethyl)cyclopentanamine hydrochloride 95%

Lieferant: Apollo Scientific

PC906755-1G PC906755-250MG
APOSPC906755-1GEA 734 EUR
APOSPC906755-1G APOSPC906755-250MG
1-(Trifluoromethyl)cyclopentanamine hydrochloride 95%
1-(Trifluoromethyl)cyclopentanamine hydrochloride
Formel: C₆H₁₀F₃N·ClH
Molecular Weight: 189,606 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143665
CAS-Nummer: 1202865-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR