2-Chloro-3,5-difluorobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC906769-1G PC906769-250MG PC906769-5G
APOSPC906769-1GEA 255 EUR
APOSPC906769-1G APOSPC906769-250MG APOSPC906769-5G
2-Chloro-3,5-difluorobenzoic acid 95%
2-Chloro-3,5-difluorobenzoic acid
Formel: C₇H₃ClF₂O₂
Molecular Weight: 192.547 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415557
CAS-Nummer: 189024-26-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR