6-Fluoro-1H-indole-3-carboxylic acid 98%

Lieferant: Apollo Scientific

PC906871-1G PC906871-250MG PC906871-5G
APOSPC906871-1GEA 144 EUR
APOSPC906871-1G APOSPC906871-250MG APOSPC906871-5G
6-Fluoro-1H-indole-3-carboxylic acid 98%
6-Fluoro-1H-indole-3-carboxylic acid
Formel: C₉H₆FNO₂
Molecular Weight: 179,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203359
CAS-Nummer: 23077-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR