4-Bromo-5-fluoro-2-methoxyphenol 95%

Lieferant: Apollo Scientific

PC907036-10G PC907036-5G
APOSPC907036-10GEA 708 EUR
APOSPC907036-10G APOSPC907036-5G
4-Bromo-5-fluoro-2-methoxyphenol 95%
4-Bromo-5-fluoro-2-methoxyphenol
Formel: C₇H₆BrFO₂
Molecular Weight: 221,024 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437599
CAS-Nummer: 886510-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR