4-(Difluoromethyl)-1-fluoro-2-nitrobenzene 95%

Lieferant: Apollo Scientific

PC907085-1G PC907085-250MG PC907085-5G
APOSPC907085-1GEA 77 EUR
APOSPC907085-1G APOSPC907085-250MG APOSPC907085-5G
4-(Difluoromethyl)-1-fluoro-2-nitrobenzene 95%
4-(Difluoromethyl)-1-fluoro-2-nitrobenzene
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698663
CAS-Nummer: 61324-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR