(2,5-Difluoro-phenyl)-methyl-amine 95%

Lieferant: Apollo Scientific

PC907124-1G PC907124-500MG
APOSPC907124-1GEA 754 EUR
APOSPC907124-1G APOSPC907124-500MG
(2,5-Difluoro-phenyl)-methyl-amine 95%
(2,5-Difluoro-phenyl)-methyl-amine
Formel: C₇H₇F₂N
Molecular Weight: 143,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11151819
CAS-Nummer: 138563-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR