(4,5-Difluoro-2-nitrophenyl)methanol 95%
Lieferant: Apollo Scientific
PC907165-1G
PC907165-250MG
APOSPC907165-1GEA
632
EUR
APOSPC907165-1G
APOSPC907165-250MG
(4,5-Difluoro-2-nitrophenyl)methanol 95%
(4,5-Difluoro-2-nitrophenyl)methanol
Formel:
C₇H₅F₂NO₃ Molecular Weight: 189,116 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20485905 CAS-Nummer: 914095-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...