Methyl 2-(2,4-difluorophenyl)acetate 95%

Lieferant: Apollo Scientific

PC907202-1G PC907202-5G
APOSPC907202-1GEA 168 EUR
APOSPC907202-1G APOSPC907202-5G
Methyl 2-(2,4-difluorophenyl)acetate 95%
Methyl 2-(2,4-difluorophenyl)acetate
Formel: C₉H₈F₂O₂
Molecular Weight: 186,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031752
CAS-Nummer: 95299-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR